Anti-B3GNT7 Rabbit Polyclonal Antibody
Catalog # PRSI26-372
Supplier: ProSci Inc.
Specifications
- Antigen name:B4GALT2
- Antigen symbol:B3GNT7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human B3GNT7.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
B3GNT7 belongs to the glycosyltransferase 31 family. It may be involved in keratane sulfate biosynthesis. B3GNT7 transfers N-acetylgalactosamine on to keratan sulfate-related glycans. It may play a role in preventing cells from migrating out of the original tissues and invading surrounding tissues.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: B3GNT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human