Order Entry
Export
ContactUsLinkComponent
Anti-MMP20 Rabbit Polyclonal Antibody
Anti-MMP20 Rabbit Polyclonal Antibody
Catalog # PRSI26-144
Supplier:  ProSci Inc.
undefined
Anti-MMP20 Rabbit Polyclonal Antibody
Catalog # PRSI26-144
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Matrix Metalloproteinase 20
  • Antigen symbol:
    MMP20
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    Enamelysin|MMP-20|MMP20|Enamel metalloproteinase|Matrix metalloproteinase-20|AI2A2
  • Immunogen:
    A synthetic peptide corresponding a region of human MMP20.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. MMP20 degrades amelogenin, the major protein component of dental enamel matrix, and so the protein is thought to play a role in tooth enamel formation. A mutation in the gene encoding MMP20, which alters the normal splice pattern and results in premature termination of the encoded protein, has been associated with amelogenesis imperfecta.Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The protein encoded by this gene degrades amelogenin, the major protein component of dental enamel matrix, and so the protein is thought to play a role in tooth enamel formation. A mutation in this gene, which alters the normal splice pattern and results in premature termination of the encoded protein, has been associated with amelogenesis imperfecta. This gene is part of a cluster of MMP genes that localizes to chromosome 11q22.3. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MMP20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human