Anti-SI Rabbit Polyclonal Antibody
PRSI26-014
- Antigen name:Sucrase-isomaltase
- Antigen symbol:SI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human SI.
- Size:50 µg
- Pk:50 µG
SI belongs to the glycosyl hydrolase 31 family. It plays an important role in the final stage of carbohydrate digestion.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human