Order Entry
Export
ContactUsLinkComponent
 
Anti-FAM19A3 Rabbit Polyclonal Antibody
  PRSI25-969
undefined
Anti-FAM19A3 Rabbit Polyclonal Antibody
  PRSI25-969

 

  • Antigen name:
    Family With Sequence Similarity 19 (chemokine (C-C motif)-like), Member A4
  • Antigen symbol:
    FAM19A3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    TAFA3|TAFA-3
  • Immunogen:
    A synthetic peptide corresponding a region of human FAM19A3.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

FAM19A3 is a member of the TAFA family which is composed of five highly homologous small secreted proteins. These proteins contain conserved cysteine residues at fixed positions, and are distantly related to MIP-1alpha, a member of the CC-chemokine family. The TAFA proteins are predominantly expressed in specific regions of the brain, and are postulated to function as brain-specific chemokines or neurokines, that act as regulators of immune and nervous cells.This gene is a member of the TAFA family which is composed of five highly homologous genes that encode small secreted proteins. These proteins contain conserved cysteine residues at fixed positions, and are distantly related to MIP-1alpha, a member of the CC-chemokine family. The TAFA proteins are predominantly expressed in specific regions of the brain, and are postulated to function as brain-specific chemokines or neurokines, that act as regulators of immune and nervous cells.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FAM19A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat