Specifications
- Antigen name:Solute Carrier Family 25 Member A16
- Antigen symbol:SLC25A16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:GDC|hML7|ML7|HGT.1|GDA|D10S105E
- Immunogen:A synthetic peptide corresponding a region of human SLC25A16.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC25A16 is a protein that contains three tandemly repeated mitochondrial carrier protein domains. The protein is localized in the inner membrane and facilitates the rapid transport and exchange of molecules between the cytosol and the mitochondrial matrix space. SLC25A16 gene has a possible role in Graves' disease.This gene encodes a protein that contains three tandemly repeated mitochondrial carrier protein domains. The encoded protein is localized in the inner membrane and facilitates the rapid transport and exchange of molecules between the cytosol and the mitochondrial matrix space. This gene has a possible role in Graves' disease.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC25A16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat