Anti-SLC44A3 Rabbit Polyclonal Antibody
Catalog # PRSI25-928
Supplier: ProSci Inc.
Specifications
- Antigen name:Solute Carrier Family 44 Member A3
- Antigen symbol:SLC44A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CTL3
- Immunogen:A synthetic peptide corresponding a region of human SLC44A3.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC44A3 is a multi-pass membrane protein. It belongs to the CTL (choline transporter-like) family. The function of the RBM34 protein remains unknown.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC44A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat