Specifications
- Antigen name:Solute Carrier Family 39 Member A4
- Antigen symbol:SLC39A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:1600025H15Rik|AWMS2|AU041686|ZIP4
- Immunogen:A synthetic peptide corresponding a region of human SLC39A4.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC39A4 is a member of the zinc/iron-regulated transporter-like protein (ZIP) family. The transmembrane protein is required for zinc uptake in the intestine. Mutations in the gene encoding SLC39A4 result in acrodermatitis enteropathica, a rare inherited defect in the absorption of dietary zinc.This gene encodes a member of the zinc/iron-regulated transporter-like protein (ZIP) family. The transmembrane protein is required for zinc uptake in the intestine. Mutations in this gene result in acrodermatitis enteropathica, a rare inherited defect in the absorption of dietary zinc. Multiple transcript variants encoding different isoforms have been found for this gene.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC39A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human