Anti-SLC25A28 Rabbit Polyclonal Antibody
Catalog # PRSI25-920
Supplier: ProSci Inc.
Specifications
- Antigen name:Solute Carrier Family 25, Member 28
- Antigen symbol:SLC25A28
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:MFRN2|MRS3/4|NPD016|MRS4L
- Immunogen:A synthetic peptide corresponding a region of human SLC25A28.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC25A28 is a mitochondrial iron transporter that mediates iron uptake. It is probably required for heme synthesis of hemoproteins and Fe-S cluster assembly in non-erythroid cells. The iron delivered into the mitochondria, presumably as Fe (2+), is then probably delivered to ferrochelatase to catalyze Fe (2+) incorporation into protoprophyrin IX to make heme.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC25A28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human