Specifications
- Antigen name:Solute Carrier Family 27 Member A4
- Antigen symbol:SLC27A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:IPS|FATP4|SLC27A4
- Immunogen:A synthetic peptide corresponding a region of human SLC27A4.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC27A4 is involved in translocation of long-chain fatty acids (LFCA) across the plasma membrane. It appears to be the principal fatty acid transporter in small intestinal enterocytes. SLC27A4 plays a role in the formation of the epidermal barrier. It is required for fat absorption in early embryogenesis. SLC27A4 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC27A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human