Order Entry
Export
ContactUsLinkComponent
Anti-SLC22A14 Rabbit Polyclonal Antibody
Anti-SLC22A14 Rabbit Polyclonal Antibody
Catalog # PRSI25-904
Supplier:  ProSci Inc.
undefined
Anti-SLC22A14 Rabbit Polyclonal Antibody
Catalog # PRSI25-904
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Solute Carrier Family 22 Member A14
  • Antigen symbol:
    SLC22A14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human SLC22A14.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

SLC22A14 is a member of the organic-cation transporter family. SLC22A14 is a transmembrane protein which is thought to transport small molecules and since this protein is conserved among several species, it is suggested to have a fundamental role in mammalian systems.This gene encodes a member of the organic-cation transporter family. It is located in a gene cluster with another member of the family, organic cation transporter like 3. The encoded protein is a transmembrane protein which is thought to transport small molecules and since this protein is conserved among several species, it is suggested to have a fundamental role in mammalian systems.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC22A14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human