Anti-SLC37A4 Rabbit Polyclonal Antibody
Catalog # PRSI25-895
Supplier: ProSci Inc.
Specifications
- Antigen name:Solute Carrier Family 37 Member A4
- Antigen symbol:SLC37A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:G6PT3|TRG-19|GSD1d|G6PT2|G6PT1|TRG19|GSD1b|PRO0685|GSD1c
- Immunogen:A synthetic peptide corresponding a region of human SLC37A4.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC37A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human