Anti-TRIM49 Rabbit Polyclonal Antibody
PRSI25-844
- Antigen name:Tripartite Motif Containing 49
- Antigen symbol:TRIM49
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:TRIM49L2|RNF18|TRIM49A
- Immunogen:A synthetic peptide corresponding a region of human TRIM49.
- Size:50 µg
- Pk:50 µG
TRIM49 contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein has been found to be preferentially expressed in testis.The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This gene has been found to be preferentially expressed in testis.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRIM49
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human