Order Entry
Export
ContactUsLinkComponent
 
Anti-SLC22A8 Rabbit Polyclonal Antibody
  PRSI25-706
undefined
Anti-SLC22A8 Rabbit Polyclonal Antibody
  PRSI25-706

 

  • Antigen name:
    Solute Carrier Family 22 Member A8
  • Antigen symbol:
    SLC22A8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    Oat3|Roct|OCT3
  • Immunogen:
    A synthetic peptide corresponding a region of human SLC22A8.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

SLC22A8 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. SLC22A8 is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney.The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC22A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human