- Antigen name:Tripartite Motif Containing 41
- Antigen symbol:TRIM41
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Tripartite motif-containing protein 41|TRIM41|E3 ubiquitin-protein ligase TRIM41|RING finger-interacting protein with C kinase|RINCK
- Immunogen:A synthetic peptide corresponding a region of human TRIM41.
- Size:50 µg
- Pk:50 µG
The TRIM41 gene encodes a protein observed in the cytoplasm and the nucleus. Nuclear transport is mediated by an N-terminal segment common to both alpha and beta isoforms, but independent of a classical nuclear localization signal sequence. TRIM41 belongs to a family of tripartite motif (TRIM) proteins defined as containing a RING finger, one or more B-box domains, and a coiled-coil region (Tanaka et al., 2005 [PubMed 16022281]). See TRIM45 (MIM 609318).
TRIM41 antibody can be used for detection of TRIM41 by ELISA at 1:312500. TRIM41 antibody can be used for detection of TRIM41 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRIM41
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human