- Antigen name:Lin-28 homolog A
- Antigen symbol:LIN28
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:RNA-binding proteináLIN-28|Zinc finger CCHC domain-containing protein 1|Lin-28 homolog A (C. elegans)|CSDD1|ZCCHC1|LIN28A
- Immunogen:A synthetic peptide corresponding a region of human LIN28.
- Size:50 µg
- Pk:50 µG
LIN28 acts as a 'translational enhancer', driving specific mRNAs to polysomes and thus increasing the efficiency of protein synthesis. The association of LIN28 with the translational machinery and target mRNAs results in an increased number of initiation events per molecule of mRNA and, indirectly, in stabilizing the mRNAs. IT binds IGF2 mRNA, MYOD1 mRNA, ARBP/36B4 ribosomal protein mRNA and its own mRNA. LIN28 is Essential for skeletal muscle differentiation program through the translational up-regulation of IGF2 expression.
LIN28 antibody can be used for detection of LIN28 by ELISA at 1:62500. LIN28 antibody can be used for detection of LIN28 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LIN28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human