Order Entry
Export
ContactUsLinkComponent
Anti-TRIM35 Rabbit Polyclonal Antibody
Anti-TRIM35 Rabbit Polyclonal Antibody
Catalog # PRSI25-205
Supplier:  ProSci Inc.
undefined
Anti-TRIM35 Rabbit Polyclonal Antibody
Catalog # PRSI25-205
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Tripartite Motif Containing 35
  • Antigen symbol:
    TRIM35
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    MAIR|HLS5
  • Immunogen:
    A synthetic peptide corresponding a region of human TRIM35.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

TRIM35 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM35 may play a role as a tumor suppressor, and is implicated in the cell death mechanism. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified.

TRIM35 antibody can be used for detection of TRIM35 by ELISA at 1:312500. TRIM35 antibody can be used for detection of TRIM35 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRIM35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human