Specifications
- Antigen name:Solute Carrier Family 35 Member D3
- Antigen symbol:SLC35D3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Antigen synonyms:Solute carrier family 35 member D3|SLC35D3|bA55K22.3|Fringe connection-like protein 1|FRCL1
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:SLC35D3 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SLC35D3.
- Tested applications:E, WB, ICC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
SLC35D3 Antibody: The solute carrier family SLC35 consists of at least 17 proteins that act as nucleotide sugar transporters localized to the Golgi apparatus and endoplasmic reticulum. The novel protein SLC35D3 is highly homologous to SLC35D1 and SLC35D2, both of which transport UDP-glucuronic acid and UDP-N-acetylgalactosamine, suggesting that SLC35D3 is also involved in the transport of nucleotide sugars. It has been suggested that SLC35D3 regulates platelet dense granules, lysosome-related organelles which contain high concentrations of several biologically important low molecular weight molecules necessary for normal blood homeostasis.
Type:
Antigen: SLC35D3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse