- Antigen name:Solute Carrier Family 35 Member D2
- Antigen symbol:SLC35D2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:UGTrel8|hfrc|HFRC1|SQV7L|HFRC|Homolog of Fringe connection protein 1|SLC35D2|UGTREL8
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:SLC35D2 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human SLC35D2.
- Tested applications:E, WB, ICC
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
SLC35D2 Antibody: The solute carrier family SLC35 consists of at least 17 proteins that act as nucleotide sugar transporters localized to the Golgi apparatus and endoplasmic reticulum. The role of the ER-resident SLC family member SLC35D2 is to transport both UDP-glucuronic acid and UDP-N-acetylgalactosamine. Its overexpression in transfected cells modulated cell surface heparin sulfate expression, suggesting that SLC35D2 is involved in heparin sulfate synthesis. SLC35D2-overexpressing cells also showed increased constitutive and hypotonic stress-stimulated release of UDP-GlcNAc, suggesting that SLC35D2 may be involved in UDP-sugar release and cell signaling.
Type:
Antigen: SLC35D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human