Order Entry
Export
ContactUsLinkComponent
Anti-DBX1 Rabbit Polyclonal Antibody
Anti-DBX1 Rabbit Polyclonal Antibody
Catalog # PRSI6455
Supplier:  ProSci Inc.
undefined
Anti-DBX1 Rabbit Polyclonal Antibody
Catalog # PRSI6455
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Developing Brain Homeobox 1
  • Antigen symbol:
    DBX1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    Developing brain homeobox protein 1|Homeobox protein DBX1|DBX1
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    DBX1 antibody was raised against a 15 amino acid synthetic peptide near the center of human DBX1.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

DBX1 Antibody: DBX1 is implicated in patterning the central nervous system during embryogenesis. DBX1 homeodomain transcription factor is expressed in progenitors which are critical in establishing cell fate allocation and cell diversity. DBX1 has a key role in regulating the distinct phenotypic features that distinguish two major classes of ventral interneurons, V0 and V1 neurons. DBX1-derived neurons are essential for the transgenic expression and generation of respiratory behavior both in vitro and in vivo.

Type:
Antigen: DBX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat