Anti-VPS39 Rabbit Polyclonal Antibody
Catalog # PRSI6183
Supplier: ProSci Inc.
Specifications
- Antigen name:Vacuolar Protein Sorting 39
- Antigen symbol:VPS39
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:TRAP1-like protein|VPS39|hVam6p|VAM6|KIAA0770|TLP
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:VPS39 antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human VPS39.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
VPS39 Antibody: VPS39 is the mammalian homolog of the yeast vacuolar protein VAM6, that in mammals, promotes lysosome clustering and fusion in vivo. VPS39 is also thought to modulate the TGF-beta response by coupling the TGF-beta receptor complex to the Smad pathway. Similar to the related protein TRAP1, the VPS39 protein is essential for early embryonic development, with mice lacking VPS39 dying at or before E6.5.
Type:
Antigen: VPS39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat