- Antigen name:Solute carrier Family 39 (zinc transporter), member 7
- Antigen symbol:ZIP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:ZIP7 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human ZIP7.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
ZIP7 Antibody: The zinc transporter ZIP7, also known as SLC39A7, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP7 was initially identified while characterizing genes in the major histocompatibility complex on chromosome 17. ZIP7 mRNA is abundantly and widely expressed and the protein localizes to the Golgi apparatus. It functions to transport intracellular zinc from the Golgi apparatus to the cytoplasm of the cell. ZIP7 expression is expressed by zinc. ZIP7 has been suggested to act a hub for tyrosine kinase activation and may thus be a potential therapeutic target for diseases such as cancer where prevention of tyrosine kinase activation would be advantageous.
Type:
Antigen: ZIP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat