Specifications
- Antigen name:Solute carrier Family 39 (zinc transporter), member 6
- Antigen symbol:ZIP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:ZIP6 antibody was raised against a 16 amino acid synthetic peptide near the center of human ZIP6.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ZIP6 Antibody: The zinc transporter ZIP6, also known as SLC39A6, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP6 was initially identified as LIV-1, an estrogen-regulated gene that has been implicated in metastatic breast cancer. Elevated ZIP6 expression has also been reported in human cervical cancer and the HeLa cell line; down-regulation of ZIP6 expression in HeLa by RNAi inhibited cell proliferation, colony formation, migration and invasiveness, as well as decreasing Snail and Slug levels, suggesting ZIP6 plays a regulatory role on the ERK1/2-Snail/Slug pathway.
Type:
Antigen: ZIP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse