Order Entry
Export
ContactUsLinkComponent
Anti-INCA1 Rabbit Polyclonal Antibody
Anti-INCA1 Rabbit Polyclonal Antibody
Catalog # PRSI5531
Supplier:  ProSci Inc.
undefined
Anti-INCA1 Rabbit Polyclonal Antibody
Catalog # PRSI5531
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Chromosome 3 Open Reading Frame 54
  • Antigen symbol:
    INCA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    INCA1 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human INCA1.
  • Tested applications:
    E, WB, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

INCA1 Antibody: INCA1 was isolated through a yeast triple hybrid approach to identify interaction partners of the cyclin A1-CDK2 heterodimer. INCA1 is evolutionarily conserved and has no homology to any known proteins. Like Cyclin A1, INCA1 is highly expressed in testis with highest levels observed at various points of testis maturation. It has been suggested that the complex of INCA1 and the cyclinA1-CDK2 dimer plays a role in several signaling pathways important for cell cycle control and meiosis. Recent experiments have shown that INCA1 also interacts with the testis protein RSB-66 and this interaction takes place in the cytoplasm.

Type:
Antigen: INCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat