Order Entry
Export
ContactUsLinkComponent
 
Anti-SLC35D1 Rabbit Polyclonal Antibody
Catalog # PRSI4649
Supplier:  ProSci Inc.
undefined
Anti-SLC35D1 Rabbit Polyclonal Antibody
Catalog # PRSI4649
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Solute Carrier Family 35 Member D1
  • Antigen symbol:
    SLC35D1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    PFTK2|ALS2CR7|UGTREL7|PFTAIRE2
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    Slc35D1 antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of the human Slc35D1.
  • Tested applications:
    E, WB
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Slc35D1 Antibody: The solute carrier family Slc35 consists of at least 17 proteins that act as nucleotide sugar transporters localized to the Golgi apparatus and endoplasmic reticulum. The role of the ER-resident Slc family member Slc35D1 is to transport both UDP-glucuronic acid and UDP-N-acetylgalactosamine. These molecules can serve as substrates for chondroitin sulfate biosynthesis and mice lacking the Slc35D1 gene developed a lethal form of skeletal dysplasia with severe shortening of limbs and facial structures. Examination of epiphyseal cartilage in these mice revealed a decreased proliferating zone with round chrondrocytes, scarce matrices, and reduced proteoglycan aggregates. Loss of function mutations in human Slc35D1 cause Schneckenbecken dysplasia, a severe skeletal dysplasia.

Type:
Antigen: SLC35D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat