Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-SARS Envelope Rabbit Polyclonal Antibody

Anti-SARS Envelope Rabbit Polyclonal Antibody

100 µg
Catalog # PRSI3531
Supplier:   ProSci Inc.
Anti-SARS Envelope Rabbit Polyclonal Antibody
Catalog # PRSI3531
Supplier:  ProSci Inc.
Anti-SARS Envelope Rabbit Polyclonal Antibody
Anti-SARS Envelope Rabbit Polyclonal Antibody

100 µg

Price
494,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

SARS Envelope Antibody: A novel coronavirus has recently been identified as the causative agent of SARS (Severe Acute Respiratory Syndrome). Coronaviruses are a major cause of upper respiratory diseases in humans. The genomes of these viruses are positive-stranded RNA approximately 27-31kb in length. SARS infection can be mediated by the binding of the viral spike protein, a glycosylated 139 kDa protein and the major surface antigen of the virus, to the angiotensin-converting enzyme 2 (ACE2) on target cells. This binding can be blocked by a soluble form of ACE2.

Type:
Antigen: SARS Envelope
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus

Documents

Specifications
  • Size:
    100 µg
  • Cat. No.:
    PRSI3531
  • Clonality:
    Polyclonal
  • Reactivity:
    Virus
  • Purification:
    Affinity chromatography purified via peptide column
  • Immunogen:
    SARS envelope antibody was raised against a synthetic peptide corresponding to amino acids at the amino-terminus of the SARS envelope protein.
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Conjugation:
    Unconjugated
  • Tested applications:
    E
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Antigen name:
    SARS Envelope
  • ELISA:
    Yes