Anti-HA Rabbit Polyclonal Antibody
Product Details & Documents
Avian Influenza Hemagglutinin Antibody: Influenza A virus is a major public health threat, killing more than 30, 000 people per year in the USA. Novel influenza virus strains caused by genetic drift and viral recombination emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals; however it is in birds that all subtypes can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. During 1997, an H5N1 avian influenza virus was determined to be the cause of death in 6 of 18 infected patients in Hong Kong. There was some evidence of human to human spread of this virus, but it is thought that the transmission efficiency was fairly low. HA interacts with cell surface proteins containing oligosaccharides with terminal sialyl residues. Virus isolated from a human infected with the H5N1 strain in 1997 could bind to oligosaccharides from human as well as avian sources, indicating its species-jumping ability.
Type:
Antigen: AVIAN INFLUENZA HEMAGGLUTININ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Documents
89415-760.pdf
Datasheet
Specifications
- Size:100 µg
- Clonality:Polyclonal
- Purification:Affinity chromatography purified via peptide column
- Isotype:IgG
- Host:Rabbit
- Antigen name:Avian Influenza Hemagglutinin
- Cat. No.:PRSI3425
- Reactivity:Virus
- Immunogen:Avian influenza hemagglutinin antibody was raised against a synthetic peptide corresponding to 14 amino acids in the middle of the avian influenza hemagglutinin protein. Efforts were made to use relatively conserved regions of the viral sequence as the an
- Conjugation:Unconjugated
- Tested applications:E
- Antigen symbol:HA
- Storage buffer:PBS containing 0.02% sodium azide
- ELISA:Yes



