- Antigen name:Tetratricopeptide Repeat Doman 5
- Antigen symbol:TTC5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:Strap|Stress-responsive activator of p300|Tetratricopeptide repeat protein 5|TTC5|TPR repeat protein 5
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:TTC5 antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human TTC5.
- Tested applications:E, WB, IHC, IF
- Purification:Immunoaffinity purified IgG
- Size:100 µg
- Pk:100 µG
TTC5 Antibody: Tetratricopeptide repeat protein 5 (TTC5) is a member of a diverse group of functionally distinct proteins that are characterized by containing one or more tetratricopeptide repeats. Each motif consists of two anti-parallel a-helices such that tandem arrays of TPR motifs generate a right-handed helical structure with an amphipathic channel that may serve to accommodate the complementary region of a target protein. While the exact function of TTC5 remains unclear, it is thought that the TPR motifs serve to mediate protein-protein interactions such as those seen with protein chaperones HSP70 and HSP90 and some proteins involved in cell stress response signaling pathways such as protein phosphatase 5, suggesting that TTC5 may also function via protein-protein interactions mediated by its TPR motifs.
Type:
Antigen: TTC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse