- Antibody type:Primary
- Antigen name:Receptor-interacting serine/threonine-protein kinase 3
- Antigen symbol:RIPK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:RIP-like protein kinase 3|Receptor-interacting protein 3|RIPK3|RIP-3|RIP3|Receptor-interacting serine/threonine-protein kinase 3
- Storage buffer:RIP3 Antibody is supplied in PBS containing 0.02% sodium azide.
- Tested applications:E, WB, IHC, IF
- Size:100 µl
- Pk:100 µG
RIP3 Antibody: Certain serine/threonine protein kinases, such as ASK1, RIP, DAP, and ZIP kinases, are mediators of apoptosis. Receptor interacting proteins including RIP and RIP2/RICK mediate apoptosis induced by TNFR1 and Fas, two prototype members in the death receptor family. A novel member in the RIP kinase family was recently identified and designated RIP3. RIP3 contains N-terminal kinase domain but, unlike RIP or RIP2, lacks the C-terminal death or CARD domain. RIP3 binds to RIP and TNFR1, mediates TNFR1 induced apoptosis, and attenuates RIP and TNFR1 induced NF-kappa B activation. Overexpression of RIP3 induces apoptosis and NF-kappa B activation. The messenger RNA of RIP3 is expressed in a subset of adult tissues.
RIP3 antibody was raised against a peptide corresponding to amino acids 473 to 486 of murine RIP3.
Type: Primary
Antigen: RIPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0