Drug and metabolite standards, levocetirizine dihydrochloride, LoGiCal®
Catalog # PROOLGCFOR1035.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:10 mg
- Standards:Reference standard
- CAS No.:130018-87-0
- Description:Levocetirizine dihydrochloride
- Formula:C₂₁H₂₇Cl₃N₂O₃
- Description:Levocetirizine dihydrochloride