Order Entry
Export
ContactUsLinkComponent
Hexadecyl palmitate 96%
Hexadecyl palmitate 96%
Certificates
Hexadecyl palmitate 96%
Certificates

Specification Test Results

  • Form :
    Solid
  • Solubility:
    Soluble in Hot Acetone

Specification Test Results

Related Information