Travoprost, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Caution: Cold Shipment required
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Caution: Cold Shipment required
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1673001.
- 1673012.
- Description:
- Travoprost
- Travoprost Related Compound A
- Synonym(s):
- (Z)-7-((1R,2R,3R,5S)-3,5-Dihydroxy-2-{(R,E)-3-hydroxy-4-[3-(trifluoromethyl)phenoxy]but-1-enyl}cyclopentyl)hept-5-enoic acid
- CAS No.:
- 157283-68-6
- 54276-17-4
- Formula:
- C₂₆H₃₅F₃O₆
- C₂₃H₂₉F₃O₆
- UN No.:
- 1648
- 1992
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1673001.TravoprostSpecifications:
More
Specifications
Cat. No.1673001.DescriptionTravoprostSynonym(s)CAS No.157283-68-6FormulaC₂₆H₃₅F₃O₆UN No.1648 - 1673012.Travoprost Related Compound ATravoprost Related Compound AUSP reference standardAmpule1,5 mLLog in for availability1 * 1,5 mL1 300,00 €Specifications:
More
Specifications
Cat. No.1673012.DescriptionTravoprost Related Compound ASynonym(s)(Z)-7-((1R,2R,3R,5S)-3,5-Dihydroxy-2-{(R,E)-3-hydroxy-4-[3-(trifluoromethyl)phenoxy]but-1-enyl}cyclopentyl)hept-5-enoic acidCAS No.54276-17-4FormulaC₂₃H₂₉F₃O₆UN No.1992



