Order Entry
Export
ContactUsLinkComponent
Tigecycline, Reference Standards
Tigecycline, Reference Standards
Supplier:  USP
Certificates
Tigecycline, Reference Standards
Supplier:  USP

Specifications

  • CAS No.:
    220620-09-7
  • UN No.:
    3077
  • Formula:
    C₂₉H₃₉N₅O₈
  • Description:
    Tigecycline
  • Synonym(s):
    (4S,4aS,5aR,12aS)-9-Amino-4,7-bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2- carboxamide hydrochloride (1:x)

Specifications