Sertraline Hydrochloride, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1612539.
- 1612528.
- 1612517.
- Description:
- Sertraline Hydrochloride
- Sertraline Hydrochloride Related Compound A
- Sertraline Hydrochloride Racemic Mixture
- Synonym(s):
- (1RS,4SR)-4-(3,4-Dichlorophenyl)-N-methyl-1,2,3,4-tetrahydro-1-naphthylamine hydrochloride
- (1RS,4RS)-4-(3,4-Dichlorophenyl)-N-methyl-1,2,3,4-tetrahydro-1-naphthylamine hydrochloride
- CAS No.:
- 79559-97-0
- 79617-89-3
- Formula:
- C₁₇H₁₈Cl₃N
- UN No.:
- 3077
- 3077
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1612539.Sertraline HydrochlorideSpecifications:
More
Specifications
Cat. No.1612539.DescriptionSertraline HydrochlorideSynonym(s)CAS No.79559-97-0FormulaC₁₇H₁₈Cl₃NUN No.3077 - 1612528.Sertraline Hydrochloride Related Compound ASertraline Hydrochloride Related Compound AUSP reference standardVial25 mgLog in for availability1 * 25 mg1 290,00 €Specifications:
More
Specifications
Cat. No.1612528.DescriptionSertraline Hydrochloride Related Compound ASynonym(s)(1RS,4SR)-4-(3,4-Dichlorophenyl)-N-methyl-1,2,3,4-tetrahydro-1-naphthylamine hydrochlorideCAS No.FormulaUN No. - 1612517.Sertraline Hydrochloride Racemic MixtureSertraline Hydrochloride Racemic MixtureUSP reference standardVial30 mgLog in for availability1 * 30 mg1 150,00 €Specifications:
More
Specifications
Cat. No.1612517.DescriptionSertraline Hydrochloride Racemic MixtureSynonym(s)(1RS,4RS)-4-(3,4-Dichlorophenyl)-N-methyl-1,2,3,4-tetrahydro-1-naphthylamine hydrochlorideCAS No.79617-89-3FormulaUN No.3077



