Order Entry
Export
ContactUsLinkComponent
Anti-IL4 Rabbit Polyclonal Antibody
Anti-IL4 Rabbit Polyclonal Antibody
Catalog # BSBTA00230
Supplier:  Boster Bio
Anti-IL4 Rabbit Polyclonal Antibody
Catalog # BSBTA00230
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    IL4
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for IL4 detection. Tested with WB, Direct ELISA in Human.

Type: Primary
Antigen: IL4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human