Anti-PTPN22 Rabbit Polyclonal Antibody
BSBTA00581-1
- Antibody type:Primary
- Antigen symbol:PTPN22
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for PTPN22 detection. Tested with WB, IHC-P, Direct ELISA in Mouse;Rat.
Type: Primary
Antigen: PTPN22
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat