Anti-PTPN1 Rabbit Polyclonal Antibody
BSBTA00613-1
- Antibody type:Primary
- Antigen name:Tyrosine-protein phosphatase non-receptor type 1
- Antigen symbol:PTPN1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Tyrosine-protein phosphatase non-receptor type 1(PTPN1) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: PTPN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat