Anti-IL22 Rabbit Polyclonal Antibody
Catalog # BSBTA00963-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Interleukin-22
- Antigen symbol:IL22
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Interleukin-22(IL22) detection. Tested with WB, ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: IL22
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat