Anti-TMEM240 Rabbit Polyclonal Antibody
Catalog # BSBTA14889
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Transmembrane protein 240
- Antigen symbol:TMEM240
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Transmembrane protein 240(TMEM240) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: TMEM240
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat