Tris(trimethylsilyl)phosphate ≥98.0% (by GC), Sigma-Aldrich®
SIAL275794-5G
: Merck
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Purity (GC):>98,0%
- Refractive index (n20/D) :1,409 (lit.)
- Colour:Colourless
- Form:Liquid
- Proton NMR spectrum :Conforms to structure
:
:314.54 g/mol
:228…229 °C (720 mmHg)) (lit.)
:3…4 °C (lit.)
:0.945 g/mL (25 °C) (lit.)
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
:10497-05-9
:C₉H₂₇O₄PSi₃
:MFCD00008267