Order Entry
Export
Orders LinkContactUsLinkComponent
Mupirocin USP, pharma grade

Mupirocin USP, pharma grade

Plastic bottle, 25 g
Catalog # A4718.0025
Supplier:   PanReac AppliChem
Mupirocin USP, pharma grade
Catalog # A4718.0025
Supplier:  PanReac AppliChem
CAS Number:  12650-69-0
Mupirocin USP, pharma grade

Plastic bottle, 25 g

CAS Number:  12650-69-0
Price
587,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Certifications: Meets the requirements of the US Pharmacopoeia (USP 40).

Synonyms:  9-(((E)-4-((2S,3R,4R,5S)-3,4-Dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoic acid
  • CAS Number:12650-69-0
  • Formula:C₂₆H₄₄O₉
  • MDL Number:MFCD01711620
  • Molecular Weight:500.629 g/mol
Specifications

Specification Test Results

  • Form:
    Solid
  • Assay (HPLC, calculated on dried substances):
    920 - 1020 µg/mg 
  • Identity:
    Passes test 
  • Crystallinity:
    Passes test 
  • pH (saturated solution; H₂O; 25 °C):
    3,5 - 4,5 
  • Water (K.F.):
    Max. 1,0% 

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g

Specifications