Order Entry
Export
ContactUsLinkComponent
Praziquantel, Reference Standards
Praziquantel, Reference Standards
Supplier:  USP
Certificates
Praziquantel, Reference Standards
Supplier:  USP

Specifications

  • CAS No.:
    54761-87-4
  • Synonym(s):
    2-benzoyl-1,2,3,6,7,11b-hexahydro-4H-pyrazino [2,1-a]isoquinolin-4-one
  • Formula:
    C₁₉H₁₈N₂O₂
  • Description:
    Praziquantel Related Compound A

Specifications