Naratriptan Related Compounds, Reference Standard
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1457469.
- 1457491.
- Description:
- Naratriptan Hydrochloride
- Naratriptan Resolution Mixture
- CAS No.:
- 143388-64-1
- Formula:
- C₁₇H₂₆ClN₃O₂S
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1457469.Naratriptan HydrochlorideNaratriptan HydrochlorideUSP reference standardVial125 mgLog in for availability1 * 125 mg512,00 €Specifications:
More
Specifications
Cat. No.1457469.DescriptionNaratriptan HydrochlorideCAS No.143388-64-1FormulaC₁₇H₂₆ClN₃O₂S - 1457491.Naratriptan Resolution MixtureNaratriptan Resolution MixtureUSP reference standardVial20 mgLog in for availability1 * 20 mg2 310,00 €Specifications:
More
Specifications
Cat. No.1457491.DescriptionNaratriptan Resolution MixtureCAS No.Formula



