Order Entry
Export
ContactUsLinkComponent
Organic reference standards, sertraline, Ph. Eur. RS
Organic reference standards, sertraline, Ph. Eur. RS
Supplier:  EDQM
Certificates
Organic reference standards, sertraline, Ph. Eur. RS
Supplier:  EDQM

Specifications

  • CAS No.:
    79559-97-0
  • Formula:
    C₁₇H₁₈Cl₃N
  • Description:
    Sertraline for peak identification

Specifications