4-Methylumbelliferyl-β-D-glucopyranoside 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18997-57-4
:242-736-7
Formula:C₁₆H₁₈O₈
MDL Number:MFCD00063694
Melting Pt:210…212 °C
:338.312 g/mol
Storage Temperature:Freezer
Specification Test Results
- Solubility:Soluble in DMF (Dimethylformamide) or water
- Purity:99%
- Physical form:Powder