Order Entry
Export
ContactUsLinkComponent
Organic reference standards, minoxidil, Ph. Eur. RS
Organic reference standards, minoxidil, Ph. Eur. RS
Supplier:  EDQM
Certificates
Organic reference standards, minoxidil, Ph. Eur. RS
Supplier:  EDQM

Specifications

  • CAS No.:
    38304-91-5
  • Formula:
    C₉H₁₅N₅O
  • Description:
    Minoxidil for system suitability

Specifications