Anti-vibration gloves, ActivArmr® 07-112
Supplier: Ansell
Certificates
About this item
The VibraGuard® gloves feature a nitrile impregnated coating on a cotton interlock/Gelfôm™ liner. They provide resistance to cuts, punctures and abrasion, as well as to grease and oil.
- Ideal for tasks involving vibrations at frequency levels greater than 300 Hz
- Engineered to reduce the vibration absorbed by workers' palm and fingers
- Help to protect from Hand-Arm Vibration Syndrome
Ideal for hand and pedestal grinders, vibrating poker, road drills, and applications including chipping hammers, heavy-duty hammers and impact rammers.
Vibration attenuation EN 10819: TRM (Transmission Ratio Medium frequency range) = 0,90/TRH (Transmission Ratio High frequency range) = 0,52
Certifications: EN 388:2016 (3221B); Cat. II
Specifications
- Catalog No:
- 112-4235
- 112-4237
- 112-4236
- Model:
- 07-112 (full-finger)
- 07-112 (full-finger)
- 07-112 (full-finger)
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Finish:
- Fully impregnated
- Fully impregnated
- Fully impregnated
- Cuff style:
- Hook & loop/Velcro® fastening
- Hook & loop/Velcro® fastening
- Hook & loop/Velcro® fastening
- EN norm:
- EN 388:2016 (3221B)
- EN 388:2016 (3221B)
- EN 388:2016 (3221B)