Cut resistant gloves, JACKSON SAFETY* G60 PURPLE NITRILE*
Supplier: KIMBERLY CLARK
Certificates
About this item
These ambidextrous gloves made from Ultra-high-molecular-weight polyethylene (HMPE) fibres provide protection against lacerations while enhancing grip and dexterity.
- Purple foam nitrile coated fingertips and flexible dotted palm for enhanced grip
- HMPE fibre enhances breathability and protection
- Knitted fibre enhances breathability and comfort
- Ambidextrous design improves cost-in-use
- Washable
- Latex-free
Typical applications include: Metal fabrication, glass handling, handling sharp parts, automotive assembly, ceramics.
Certifications: EN 388 (434X); Cat. II
Specifications
- Catalog No:
- 112-4859
- 112-4860
- 112-1236
- 112-4858
- 112-4861
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Finish:
- Foam nitrile coated fingertips, dotted palm
- Foam nitrile coated fingertips, dotted palm
- Foam nitrile coated fingertips, dotted palm
- Foam nitrile coated fingertips, dotted palm
- Foam nitrile coated fingertips, dotted palm
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (434X)
- EN 388 (434X)
- EN 388 (434X)
- EN 388 (434X)
- EN 388 (434X)