Anti-IgM (µ chain) Rabbit Polyclonal Antibody (Biotin)
Catalog # 30-8075-M001
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin M mu
- Antigen symbol:Human IgM (µ chain)
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Epitope:µ chain
- Storage temperature:Store at 2-8 °C
- Pk:1 mg
Specifications
About this item
Rabbit anti-Human IgM (mu chain) - Affinity Pure, Biotin Conjugate
Type: Secondary
Antigen: Human IgM (µ chain)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ chain
Host: Rabbit
Isotype:
Reactivity: Human