Anti-IgM Goat Polyclonal Antibody
Catalog # 70-8064-M002
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin M
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western blot:Yes
- Epitope:Heavy and light chain
- Storage temperature:Store at 2-8 °C
- Pk:2 mg
Specifications
About this item
Goat anti-Mouse IgM (H&L) - Affinity Pure
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse