Anti-IgM Goat Polyclonal Antibody (Biotin)
Catalog # 30-8028-M001
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin M mu
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Epitope:µ chain
- Storage temperature:Store at 2-8 °C
- Pk:1 mg
Specifications
About this item
Goat anti-Human IgM (mu chain) - Affinity Pure, Biotin Conjugate
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Human